Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: Bile Esculin Azide Agar (Enterococcosel™)


Modified esculin bile agar, for the rapid, selective detection and enumeration of enterococci

Manufacturer: bd  B221492

 View more versions of this product

Catalog No. B21492X

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Yellow Brown
For Use With (Application) Enterococci
Format Liquid
Manufacturing Origin United States
Media Format Pancreatic digest of Casein 17g/L, Peptic digest of Animal Tissue 3g/L, Yeast extract 5g/L, Oxgall 10g/L, Sodium Chloride 5g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Azide 0.25g/L, Sodium Citrate 1g/L, Agar 13.5g/L
Media Type Bile Esculin Azide Agar
Packaging Plate
Quantity 20/Pk.
Shelf Life 112 Days
Storage Requirements 2-25°C
Accessories

112 Days