Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Media: BCYE Selective Agar


For isolation of Legionella species from clinical and nonclinical specimens

Manufacturer: bd  B297880

 View more versions of this product

Catalog No. L97880

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Green
For Use With (Application) Legionella species from contaminated clinical specimens (with vancomycin)
Format Liquid
Manufacturing Origin United States
Media Format BCYE Agar 38.65g/L, Polymyxin B 40000U, Anisomycin 80mg/L, Vancomycin 0.5mg/L
Media Type BCYE selective Agar with PAV
Packaging Plate
Quantity 10/Pk.
Shelf Life 98 Days
Storage Requirements 2-8°C
Accessories

98 Days