Please login to your online account to display your discounted pricing

BD BBL™ Prepared Plated Chromogenic Media: Modified mTEC Agar


Culture media tests water for the presence of E.coli with membrane filtration method

Manufacturer: bd  B215044

 View more versions of this product

Catalog No. B15044

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Quantity 20/Pk.
For Use With (Application) The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique
Media Type Modified m TEC Agar
Packaging Plate
Shelf Life 84 Days
Format Liquid
Manufacturing Origin United States
Media Format No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 10g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, 5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide 0.5g/L, Agar 15g/L
Certifications/Compliance US, EPA