missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Chromogenic Media: Modified mTEC Agar
Culture media tests water for the presence of E.coli with membrane filtration method
$229.39 - $2,159.19
Specifications
Certifications/Compliance | US, EPA |
---|---|
Format | Liquid |
For Use With (Application) | The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique |
Media Type | Modified m TEC Agar |
Packaging | Plate |
Description
Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.
- Any colonies present will exhibit red or magenta color within 24 hours of the test
- Conforms to EPA Method 1603
Order Info
Available in prepared 15 × 60mm plates.Note: Minimum order quantity of 5 is required for purchase.
Specifications
US, EPA | |
The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique | |
Plate | |
No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 10g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, 5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide 0.5g/L, Agar 15g/L |
Liquid | |
Modified m TEC Agar | |
United States | |
84 Days |