missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Prepared Plated Chromogenic Media: m-EI Agar Base
Culture media tests water for the presence of Enterococci by membrane filtration method
$2,466.54 - $2,466.54
Specifications
Certifications/Compliance | US, EPA |
---|---|
Format | Liquid |
For Use With (Application) | The chromogenic detection and enumeration of Enterococci in Water by the single-step membrane filtration technique |
Media Type | m EI Agar |
Packaging | Plate |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B15047
|
BD
B215047 |
100/Pk. |
Pack of 100 Each for $2,466.54
|
|
Description
- Any colonies present will exhibit blue halo within 24 hours of the test
- Conforms to EPA Method 1600
Order Info
Available in prepared 15 x 60mm plates.
Specifications
US, EPA | |
The chromogenic detection and enumeration of Enterococci in Water by the single-step membrane filtration technique | |
Plate | |
Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L, Nalidixic Acid 0.24g/L, Triphenyltetrazolium Chloride 0.02g/L | |
2-8°C |
Liquid | |
m EI Agar | |
United States | |
70 Days |