missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Mycoflask™ Simmons Citrate Agar Prepared Media Slants
Used for differentiation of gram-negative bacteria on the basis of citrate utilization
Supplier: BD 221026
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, AOAC, BAM, CCAM, COMPF, ISO | |
2°C to 8°C | |
United States | |
Ammonium Dihydrogen Phosphate 1g/L, Dipotassium Phosphate 1g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Magnesium Sulfate 0.2g/L, Agar 15g/L, Bromthymol blue 0.08g/L | |
10/Pk. |
Green | |
Liquid | |
Simmons Citrate Agar | |
Tube with Tight Cap |
For in vitro diagnostic use