missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Mycoflask™ Selenite-F Broth Prepared Media

For isolation of Salmonella from feces, urine, water, foods and other materials of sanitary importance

$22.74 - $22.74

Specifications

Certifications/Compliance CE, BS12, MCM9, SMWW
Color Beige
Format Liquid
For Use With (Application) Isolation of Salmonella from feces, urine, Water, foods and other materials of sanitary importance
Packaging Tube with Tight Cap
View More Specs

Products 1
Catalog Number Mfr. No. Media Type Quantity Price Quantity  
Catalog Number Mfr. No. Media Type Quantity Price Quantity  
221020 View Documents BD
B221020
Selenite-F Broth 10 Pk. Pack of 10 Each for $22.74
 
Description

Description

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications

Specifications

CE, BS12, MCM9, SMWW
Liquid
Tube with Tight Cap
Pancreatic digest of Casein 5g/L, Lactose 4g/L, Sodium Selenite 4g/L, Sodium Phosphate 10g/L
Beige
Isolation of Salmonella from feces, urine, Water, foods and other materials of sanitary importance
United States
2° to 8°C

For in vitro diagnostic use.