Please login to your online account to display your discounted pricing

BD BBL™ Mycoflask™ OF (Oxidation Fermentation) Media


Determines oxidative and fermentative metabolism of carbohydrates by Gram-negative rods on the basis of acid reaction

Manufacturer: bd  221338

 View more versions of this product

 Disclaimers

For in vitro diagnostic use.

Catalog No. BB21338

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Storage Requirements 2-8°C
Color Green
For Use With (Application) Determination of oxidative and fermentative metabolism of carbohydrates by gram negative rods on the basis of Acid reaction
Format Liquid
Manufacturing Origin United States
Media Format Pancreatic digest of Casein 2g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 0.3g/L, Agar 2.5g/L, Bromthymol blue 0.03g/L
Media Type OF media with Xylose
Packaging Tube
Shelf Life 365 Days
Certifications/Compliance ISO/DIS, 11014