missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ Mycoflask™ Mueller Hinton II Broth (Cation-Adjusted) Prepared Media

For use in quantitative procedures for susceptibility testing of aerobic and facultatively anaerobic bacteria from clinical specimens

Manufacturer:  BD B298268

 View more versions of this product

Catalog No. L98268



Description

Description

  • Formulated to have low thymine and thymidine content
  • Consisting of microorganisms, nutrient medium and antimicrobial agent

Specifications

Specifications

ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9
Liquid
Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L
Tube with Tight Cap
2° to 25°C
Clear
United States
Mueller Hinton II Broth
5mL
Documents

For in vitro Diagnostic Use.