Please login to your online account to display your discounted pricing

BD BBL™ Middlebrook OADC Enrichment

Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.

$253.77 - $620.44

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Viscous
For Use With (Application) Dehydrated culture media
Shelf Life 365 Days
Storage Requirements 2-8°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B11886

 
bd
B211886
Tube 20mL Case of 10 Each for $253.77

B12351

 
bd
B212351
Bottle 500mL Each for $620.44
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Viscous
For Use With (Application) Dehydrated culture media
Shelf Life 365 Days
Storage Requirements 2-8°C
Additive Middlebrook OADC Enrichment

  • Dehydrated format
  • Culture media
  • DNR (Latex) free