missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Test Kits

Offers superior detection time in a user-friendly, fluorescent test format

$1,185.95 - $1,185.95

Specifications

Certifications/Compliance CE
Product Type Test Kit (Prefilled with Indicator and Broth)
For Use With (Application) For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
Storage Requirements 2 to 25°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4345111 BD
B245111
25 tubes Pack of 25 Each for $1,185.95
Specifications

Description

  • Creates an environment suitable for rapid mycobacterial growth
  • Recommended for testing sputum, stool specimens, body fluids and tissue
  • The tube uses fluorometric technology to aid in the reading of results
  • Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
  • The Mycobacteria can be removed from the tube for further testing
  • To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
  • Tubes fit into standard procedures for decontamination and waste disposal

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Description & Specifications

Specifications

CE
Test Kit (Prefilled with Indicator and Broth)
For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
2 to 25°C