Please login to your online account to display your discounted pricing

BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Reagents


Offers superior detection time in a user-friendly, fluorescent test format

Manufacturer: bd  B245116

 View more versions of this product

Catalog No. B4345116

  • / Pack of 6 Each
 

Description & Specifications

Specifications

For Use With (Application) For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
Storage Requirements 2 to 25°C
Quantity 6 vials
Volume (Metric) 6 Vials
Certifications/Compliance CE
Product Type OADC Enrichment