Please login to your online account to display your discounted pricing

BD BBL™ Medium Herrold's Egg Yolk Agar: Without Mycobactin J

Selective isolation and differentiation of Mycobacterium paratuberculosis

$174.79 - $174.79

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Green
Format Solid
For Use With (Application) Selective isolation and differentiation of Mycobacterium paratuberculosis
Packaging Slant
View More Specs
Products
Catalog Number Mfr. No. Media Type Quantity Price Quantity    

B222240

 
bd
B222240
Herrold's Egg Yolk Agar with Amphotericin, Nalidixic Acid, Vancomycin, without Mycobactin J 10/Pk. Pack of 10 Each for $174.79
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Green
Format Solid
For Use With (Application) Selective isolation and differentiation of Mycobacterium paratuberculosis
Packaging Slant
Manufacturing Origin United States
Shelf Life 365 Days

  • Available with amphotericin, nalidixic acid, vancomycin
  • Features “Slant Surface”, a widely used format that gives enough surface area for growth and visualization

Safety and Handling

365 Days