Please login to your online account to display your discounted pricing

BD BBL™ Medium Herrold's Egg Yolk Agar: Without Mycobactin J


Selective isolation and differentiation of Mycobacterium paratuberculosis

Manufacturer: bd  B222240

 View more versions of this product

Catalog No. B222240

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Packaging Slant
Color Light Green
Manufacturing Origin United States
Media Type Herrold's Egg Yolk Agar with Amphotericin, Nalidixic Acid, Vancomycin, without Mycobactin J
Certifications/Compliance ISO/DIS, 11014
Format Solid
Shelf Life 365 Days
For Use With (Application) Selective isolation and differentiation of Mycobacterium paratuberculosis
Media Format Pancreatic digest of Casein 9g/L, Beef extract 2.7g/L, Sodium Chloride 4g/L, Sodium Pyruvate 4.1g/L, Malachite green 0.1g/L, Amphotericin B 0.05g/L, Agar 15.3g/L, Nalidixic Acid 0.05g/L, Vancomycin 0.05g/L, Egg Yolks 100mL, Glycerol 27mL, Purified Water 873.0mL
Accessories

365 Days