Please login to your online account to display your discounted pricing

BD BBL™ Fluid Thioglycollate Dehydrated Culture Media


Used for sterility testing of viscous materials

Manufacturer: bd  B211260

 View more versions of this product

Catalog No. B11260

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophile
Format Dehydrated
Media Format Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Media Type Thioglycollate Medium Fluid
Packaging Bottle
Product Type Thioglycollate medium fluid
Quantity 500g
Shelf Life 1460 Days
Storage Requirements 15-35°C
Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, COMPF, EP, USDA, USP