Please login to your online account to display your discounted pricing

BD BBL™ Fluid Thioglycollate Dehydrated Culture Media

Used for sterility testing of viscous materials

$149.85 - $591.72

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, COMPF, EP, USDA, USP
Format Dehydrated
For Use With (Application) For sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophile
Media Format Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Shelf Life 1460 Days
View More Specs
Products
Catalog Number Mfr. No. Media Type Packaging Quantity Price Quantity    

B11260

 
bd
B211260
Thioglycollate Medium Fluid Bottle 500g Each for $149.85

B11263

 
bd
B211263
Thioglycollate Medium Pail 2270g Each for $591.72
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, COMPF, EP, USDA, USP
Format Dehydrated
For Use With (Application) For sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophile
Media Format Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Shelf Life 1460 Days
Storage Requirements 15-35°C