Please login to your online account to display your discounted pricing

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Proteose Peptone


Enzymatic digest of protein, high in proteoses

Manufacturer: bd  B211684

 View more versions of this product

Catalog No. DF0120-17-6

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Format Dehydrated
Quantity 500g
Shelf Life 1825 Days
With Additives Proteose Peptone
Certifications/Compliance LUO, MUO
Packaging Type Bottle
Packaging Bottle
Storage Requirements <30°C
Quantity 500g