Please login to your online account to display your discounted pricing

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline


Fluorescent Antibody reagents

Manufacturer: bd  B223143

 View more versions of this product

Catalog No. DF2314-33-8

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Storage Requirements <30°C
Quantity 10g
Shelf Life 1825 Days
Packaging Vial
Format Dehydrated
Certifications/Compliance ISO/DIS, 11014
For Use With (Application) Fluorescent Antibody reagents
Additive Phosphate Buffered Saline
Accessories

1825 Days