Please login to your online account to display your discounted pricing

BD BBL™/Difco™ Bacto™ Media Additives/Ingredients/Reagents: Peptone


Enzymatic digest of protein

Manufacturer: bd  B211820

 View more versions of this product

Catalog No. DF0118-07-2

  • / Each
 

Description & Specifications

Specifications

Quantity 2kg
Storage Requirements 2-25°C
With Additives Peptone
Certifications/Compliance LUO, MUO
Packaging Type Pail
Shelf Life 1825 Days
Format Powder
Packaging Pail
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Quantity 2000g

  • Contains only negligible quantity of proteoses and more complex constituents
  • Used as organic nitrogen source in microbiological culture media for cultivation of a variety of bacteria and fungi

Accessories

1825 Days