missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD BBL™/Difco™ Media Additives/Ingredients/Reagents

Use with dehydrated culture media

$2,150.62 - $2,150.62

Specifications

Additive Trypsin
Format Powder
For Use With (Application) Dehydrated culture media
Packaging Bottle
Storage Requirements <30°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Certifications/Compliance Quantity Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Quantity Price Quantity  
D015217 BD
B215250
CE 500g Each for $2,150.62
Specifications Description & Specifications

Specifications

Trypsin
Dehydrated culture media
<30°C
Powder
Bottle