Please login to your online account to display your discounted pricing

BD BBL™/Difco™ Media Additives/Ingredients/Reagents


Use with dehydrated culture media

Manufacturer: bd  B215250

 View more versions of this product

Catalog No. D015217

  • / Each
 

Description & Specifications

Specifications

Quantity 500g
Format Powder
Storage Requirements <30°C
Packaging Bottle
Certifications/Compliance CE
Shelf Life 1825 Days
Additive Trypsin
For Use With (Application) Dehydrated culture media
Accessories

1825 Days