Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Tryptone Glucose Yeast Agar (Standard Methods Agar)


For obtaining microbial plate counts from milk and dairy products, foods, water and other materials of sanitary importance

Manufacturer: bd  B212455

 View more versions of this product

Catalog No. B12455

  • / Each
 

Description & Specifications

Specifications

Packaging Bottle
Format Dehydrated
Product Type Plate Count Agar
For Use With (Application) For obtaining microbial plate counts from Milk and dairy product, food Water and other materials of sanitary importance
Storage Requirements Refrigerate
Media Format Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L
Media Type Standard Methods Agar (Tryptone Glucose Yeast Agar)
Quantity 100g
Certifications/Compliance CE, AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW, USDA
Shelf Life 1460 Days
Accessories

1460 Days