Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Trypticase Soy Broth (Soybean-Casein Digest Broth)


Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Manufacturer: bd  B211771

 View more versions of this product

Catalog No. B11771

  • / Each
 

Description & Specifications

Specifications

Product Type Trypticase™ Soy broth
Packaging Pail
Quantity 5 lb.
Media Type Trypticase Soy Broth
Storage Requirements 2-25°C
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Certifications/Compliance AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP
Format Dehydrated
Shelf Life 1460 Days
Media Format Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
Accessories

1460 Days