missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Dehydrated Culture Media: Soybean Casein Digest Agar
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Supplier: BD 211047
Description
Applications:
Isolation of non-fastidious and fastidious
Specifications
ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Dehydrated | |
Agar | |
Isolation of non-fastidious and fastidious | |
11.3 kg |
25°C | |
Soy Agar | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Pail |