Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Soybean Casein Digest Agar


Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Manufacturer: bd  B211046

 View more versions of this product

Catalog No. B11046

  • / Each
 

Description & Specifications

Specifications

Format Dehydrated
Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Shelf Life 1460 Days
Packaging Pail
Quantity 2.3kg
Product Type Soybean Casein digest Agar
Storage Requirements 25°C
For Use With (Application) Isolation of non-fastidious and fastidious
Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
Media Type Soy Agar
Accessories

1460 Days