Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80


Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Manufacturer: bd  B211764

 View more versions of this product

Catalog No. B11764

  • / Each
 

Description & Specifications

Specifications

Format Dehydrated
Product Type Soy Agar with Lecithin and Polysorbate 80
Certifications/Compliance CE, CCAM
Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
Shelf Life 1460 Days
Packaging Bottle
Quantity 500g
Media Type Soy Agar with Lecithin and Polysorbate 80
For Use With (Application) For Aerobic Plate count (Microbial Limit Test) for Water-miscible cosmetic product containing preservatives
Accessories

1460 Days