Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

$190.87 - $696.31

Specifications

Certifications/Compliance CCAM
Format Dehydrated
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Product Type Soy Agar with Lecithin and Polysorbate 80
Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/mL, Polysorbate 80 5g/L, Agar 15g/L
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B11764

 
bd
B211764
Bottle 500g Each for $190.87

B12263

 
bd
B212263
Pail 5 lb. Each for $696.31
Description & Specifications

Specifications

Certifications/Compliance CCAM
Format Dehydrated
For Use With (Application) Detection and enumeration of microorganisms present on surfaces of sanitary importance
Product Type Soy Agar with Lecithin and Polysorbate 80
Media Format Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/mL, Polysorbate 80 5g/L, Agar 15g/L
Shelf Life 1460 Days

Detection of microorganisms on surfaces sanitized with quaternary ammonium compounds

CE, CCAM