Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Salmonella Shigella Agar (SS Agar)

For selective isolation of Salmonella and some Shigella species

$171.10 - $171.10

Specifications

Format Dehydrated
Media Type Salmonella Shigella Agar
Packaging Bottle
Product Type Salmonella Shigella Agar
Media Format Beef extract 5g/L, Pancreatic digest of Casein 2.5g/L, Peptic digest of Animal Tissue 2.5g/L, Lactose 10g/L, Bile Salts 8.5g/L, Sodium Citrate 8.5g/L, Sodium Thiosulfate 8.5g/L, Ferric Citrate 1g/L, Agar 13.5g/L, Brilliant green 0.33mg/L, Neutral red 25mg/L
View More Specs
Products
Catalog Number Mfr. No. Media Format For Use With (Application) Packaging Product Type Price Quantity    

B11597

 
bd
B211597
Beef extract 5g/L, Pancreatic digest of Casein 2.5g/L, Peptic digest of Animal Tissue 2.5g/L, Lactose 10g/L, Bile Salts 8.5g/L, Sodium Citrate 8.5g/L, Sodium Thiosulfate 8.5g/L, Ferric Citrate 1g/L, Agar 13.5g/L, Brilliant green 0.33mg/L, Neutral red 25mg/L Isolation of pathogenic enteric bacilli, especially those belonging to the genus Salmonella Bottle Salmonella Shigella Agar Each for $171.10
Description & Specifications

Specifications

Format Dehydrated
Media Type Salmonella Shigella Agar
Packaging Bottle
Product Type Salmonella Shigella Agar
Media Format Beef extract 5g/L, Pancreatic digest of Casein 2.5g/L, Peptic digest of Animal Tissue 2.5g/L, Lactose 10g/L, Bile Salts 8.5g/L, Sodium Citrate 8.5g/L, Sodium Thiosulfate 8.5g/L, Ferric Citrate 1g/L, Agar 13.5g/L, Brilliant green 0.33mg/L, Neutral red 25mg/L
Shelf Life 1460 Days