missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ Dehydrated Culture Media: Salmonella Shigella Agar (SS Agar)
For selective isolation of Salmonella and some Shigella species
Supplier: BD 211597
Specifications
CE, BS12, CMPH2, COMPF, MCM9 | |
Dehydrated | |
Agar | |
Isolation of pathogenic enteric bacilli, especially those belonging to the genus Salmonella | |
500 g |
Refrigerate | |
Salmonella Shigella Agar | |
Beef extract 5g/L, Pancreatic digest of Casein 2.5g/L, Peptic digest of Animal Tissue 2.5g/L, Lactose 10g/L, Bile Salts 8.5g/L, Sodium Citrate 8.5g/L, Sodium Thiosulfate 8.5g/L, Ferric Citrate 1g/L, Agar 13.5g/L, Brilliant green 0.33mg/L, Neutral red 25mg/L | |
Bottle |