Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Sabouraud Dextrose Agar

For cultivation of yeasts, molds and aciduric microorganisms

$134.77 - $548.56

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Format Dehydrated
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Media Type Sabouraud Dextrose Agar
Product Type Sabouraud Dextrose Agar
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B11585

 
bd
B211585
Pail 2kg Each for $548.56

B11584

 
bd
B211584
Bottle 500g Each for $134.77
Description & Specifications

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Format Dehydrated
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Media Type Sabouraud Dextrose Agar
Product Type Sabouraud Dextrose Agar
Media Format Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
Shelf Life 1460 Days
Storage Requirements Refrigerate
Safety and Handling

1460 Days