Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Middlebrook ADC Enrichment


Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook ADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes

Manufacturer: bd  B211887

 View more versions of this product

Catalog No. B11887

  • / Case of 10 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Packaging Tube
Additive Middlebrook ADC
Shelf Life 243 Days
For Use With (Application) Dehydrated culture media
Format Liquid
Quantity 20mL
Storage Requirements 2-8°C

  • Dehydrated format
  • Culture media
  • DNR (Latex) free

Accessories

243 Days