Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Middlebrook ADC Enrichment


Used as a growth supplement for microbiology research.  BD Difco™ BBL™ Middlebrook ADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes

Manufacturer: bd  B212352

 View more versions of this product

Catalog No. B12352

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Packaging Bottle
Quantity 100mL
Additive Middlebrook ADC
Format Liquid
Certifications/Compliance ISO/DIS, 11014
Storage Requirements 2-8°C
For Use With (Application) Dehydrated culture media
Shelf Life 243 Days

  • Dehydrated format
  • Culture media
  • DNR (Latex) free

Accessories

243 Days