Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: MacConkey Agar


For differential isolation of enteric bacilli based on lactose fermentation

Manufacturer: bd  B211390

 View more versions of this product

Catalog No. B11390

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-30°C
Product Type MacConkey Agar
Media Format Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
Quantity 2.27kg
Shelf Life 1460 Days
For Use With (Application) For isolating and differentiating lactose-fermenting from lactose-nonfermenting gram negative enteric Bacilli
Media Type MacConkey Agar
Packaging Pail
Format Dehydrated
Certifications/Compliance AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Accessories

1460 Days