Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Lauryl Sulfate Broth


For detection of coliform bacteria

Manufacturer: bd  B211339

 View more versions of this product

Catalog No. B11339

  • / Each
 

Description & Specifications

Specifications

Quantity 2.27kg
Format Dehydrated
Product Type Lauryl Sulfate broth
Certifications/Compliance AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW
Media Format Pancreatic digest of Casein 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L
Shelf Life 1460 Days
For Use With (Application) Detection of Coliform bacteria
Packaging Pail
Media Type Lauryl Sulfate Broth
Storage Requirements Refrigerate
Accessories

1460 Days