Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Lauryl Sulfate Broth

For detection of coliform bacteria

$149.87 - $566.92

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW
Format Dehydrated
For Use With (Application) Detection of Coliform bacteria
Media Type Lauryl Sulfate Broth
Product Type Lauryl Sulfate broth
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B11338

 
bd
B211338
Bottle 500g Each for $149.87

B11339

 
bd
B211339
Pail 2.27kg Each for $566.92
Description & Specifications

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW
Format Dehydrated
For Use With (Application) Detection of Coliform bacteria
Media Type Lauryl Sulfate Broth
Product Type Lauryl Sulfate broth
Media Format Pancreatic digest of Casein 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L
Shelf Life 1460 Days
Storage Requirements Refrigerate

AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW