Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Columbia CNA Agar

For selective isolation of gram positive cocci

$345.92 - $1,093.28

Specifications

Format Dehydrated
For Use With (Application) For selective isolation of gram positive cocci
Media Type Columbia CNA Agar
Product Type Columbia CNA Agar
Media Format Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B12104

 
bd
B212104
Bottle 500g Each for $345.92

B94221X

 
bd
B294221
Pail 2.26kg Each for $1,093.28
Description & Specifications

Specifications

Format Dehydrated
For Use With (Application) For selective isolation of gram positive cocci
Media Type Columbia CNA Agar
Product Type Columbia CNA Agar
Media Format Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L
Shelf Life 1460 Days
Storage Requirements 2-8°C