Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Columbia CNA Agar


For selective isolation of gram positive cocci

Manufacturer: bd  B294221

 View more versions of this product

Catalog No. B94221X

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-8°C
Product Type Columbia CNA Agar
Quantity 2.26kg
Packaging Pail
Format Dehydrated
Media Type Columbia CNA Agar
Media Format Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L
Shelf Life 1460 Days
For Use With (Application) For selective isolation of gram positive cocci
Accessories

1460 Days