Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Buffered Peptone Water


Used for preenriching injured Salmonella species from food specimens to increase recovery

Manufacturer: bd  B212367

 View more versions of this product

Catalog No. B12367

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cream-white to Light Tan
Format Dehydrated
Packaging Bottle
Product Type Buffered Peptone Water
Quantity 500g
Shelf Life 1460 Days
Storage Requirements Refrigerate
Media Format Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
Media Type Buffered Peptone Water
Certifications/Compliance BAM, CCAM, ISO, USDA