Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Buffered Peptone Water


Used for preenriching injured Salmonella species from food specimens to increase recovery

Manufacturer: bd  B212345

 View more versions of this product

Catalog No. B12345

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For pre-enriching injured Salmonella species from food specimens to increase recovery
Format Dehydrated
Media Format Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
Media Type Buffered Peptone Water
Packaging Pail
Product Type Buffered Peptone Water
Quantity 2270g
Shelf Life 1460 Days
Certifications/Compliance BAM, CCAM, ISO, USDA