Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Buffered Peptone Water

Used for preenriching injured Salmonella species from food specimens to increase recovery

$234.72 - $395.67

Specifications

Certifications/Compliance BAM, CCAM, ISO, USDA
Format Dehydrated
Media Type Buffered Peptone Water
Product Type Buffered Peptone Water
Media Format Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Packaging Quantity Price Quantity    

B12345

 
bd
B212345
For pre-enriching injured Salmonella species from food specimens to increase recovery Pail 2270g Each for $395.67

B12367

 
bd
B212367
Cream-white to Light Tan Bottle 500g Each for $234.72
Description & Specifications

Specifications

Certifications/Compliance BAM, CCAM, ISO, USDA
Format Dehydrated
Media Type Buffered Peptone Water
Product Type Buffered Peptone Water
Media Format Peptone 10g/L, Sodium Chloride 5g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 1.5g/L
Shelf Life 1460 Days

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

BAM, CCAM, ISO, USDA