Please login to your online account to display your discounted pricing

BD BBL™ Dehydrated Culture Media: Brain Heart Infusion Agar

Use for isolation and cultivation of wide variety of fungal species, icluding bacteria, yeast and molds

$241.21 - $241.21

Specifications

Format Dehydrated
For Use With (Application) For bacteria, Yeasts, molds and fungal species
Media Type BHI Agar
Product Type Brain heart infusion Agar
Media Format Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Gelatin 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B11065

 
bd
B211065
Bottle 500g Each for $241.21
Description & Specifications

Specifications

Format Dehydrated
For Use With (Application) For bacteria, Yeasts, molds and fungal species
Media Type BHI Agar
Product Type Brain heart infusion Agar
Media Format Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Gelatin 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L

  • Formulation: Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Gelatin 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L

AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA

Safety and Handling

1460 Days