missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ D/E Neutralizing Agar Sterile Pack Media - Rodac™ Plates
Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure
$77.60 - $77.60
Specifications
Color | Purple |
---|---|
Format | Rodac |
Packaging | Plate |
Manufacturing Origin | United States |
Shelf Life | 112 Days |
Catalog Number | Mfr. No. | Certifications/Compliance | Media Type | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Media Type | Quantity | Price | Quantity | |||
B4321232
|
BD
B221232 |
USP | D/E neutralizing Agar | 10/Pk. |
Pack of 10 Each for $77.60
|
|
Description
Rodac (Replicate Organism Detection and Counting) plates for surface sampling.
To use:
- Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
- When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
- Use the included inner bag to transport plates from your critical environment
Compliance
Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.
Specifications
Purple | |
Plate | |
112 Days |
Rodac | |
United States |