missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD BBL™ D/E Neutralizing Agar Isolator Pack Media - Rodac™ Plates
Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure
$1,015.97 - $1,015.97
Specifications
Certifications/Compliance | AAMI |
---|---|
Color | Gray |
Format | Liquid |
Media Type | D/E Neutralizing Agar |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Packaging | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging | Quantity | Price | Quantity | ||||
B292646
|
BD
B292646 |
Sterile Rodac™ Plate | 100/Pk. |
Pack of 100 Each for $1,015.97
|
|
Description
Rodac (Replicate Organism Detection and Counting) plates for surface sampling.
To use:
- Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
- When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
- Use the included inner bag to transport plates from your critical environment
Compliance
Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.
Specifications
AAMI | |
Liquid | |
United States | |
2-8°C |
Gray | |
D/E Neutralizing Agar | |
112 Days |