Please login to your online account to display your discounted pricing

BD BBL™ D/E Neutralizing Agar Isolator Pack Media - Rodac™ Plates


Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Manufacturer: bd  B292645

 View more versions of this product

Catalog No. B292645

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Format Rodac
Quantity 10/Pk.
Sterile Yes
Storage Requirements 2-8°C
Color Gray
For Use With (Application) Monitoring surfaces and air in Isolator system, clean rooms and other environmentally controlled areas when sterility of the medium is of importance
Includes Inner bag
Manufacturing Origin United States
Media Format Pancreatic digest of Casein 5g/L, Yeast extract 5g/L, Dextrose 9g/L, Sodium Bisulfite 2.5g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Lecithin 7g/L, Polysorbate 80 5g/L, Bromcresol Purple 0.02g/L, Agar 15g/L
Media Type D/E neutralizing Agar
Packaging Plate
Shelf Life 112 Days
Weight (Metric) 900g
Certifications/Compliance AAMI