Please login to your online account to display your discounted pricing

BD BBL™ CTA Medium


Simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms; addition of carboyhydrates allows use in the determination of fermentation reactions

Manufacturer: bd  B221645

 View more versions of this product

Catalog No. BB21645

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Format Liquid
Packaging Tube
Media Type CTA Culture Medium with Sucrose
Color Beige
Media Format L-Cystine 0.5g/L, Pancreatic digest of Casein 20g/L, Agar 2.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Phenol red 0.017g/L
Storage Requirements 2° to 8°C
Certifications/Compliance ISO/DIS, 11014, AOAC
Quantity 8mL
Manufacturing Origin United States
Shelf Life 548 Days
For Use With (Application) For maintaining the growth of microorganisms
Accessories

548 Days