Please login to your online account to display your discounted pricing

BD BBL™ CTA Medium


Simple, basic medium for the maintenance and detection of motility of a wide variety of microorganisms; addition of carboyhydrates allows use in the determination of fermentation reactions

Manufacturer: bd  B221633

 View more versions of this product

Catalog No. BB21633

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Color Beige
For Use With (Application) For maintaining the growth of microorganisms
Format Liquid
Manufacturing Origin United States
Media Format L-Cystine 0.5g/L, Pancreatic digest of Casein 20g/L, Agar 2.5g/L, Sodium Chloride 5g/L, Sodium Sulfite 0.5g/L, Phenol red 0.017g/L
Media Type CTA Culture Medium with Dextrose
Packaging Tube
Quantity 8mL
Shelf Life 548 Days
Storage Requirements 2° to 8°C
Certifications/Compliance CE, AOAC