Please login to your online account to display your discounted pricing

BD Bacto™ Dehydrated Culture Media: Tryptic Soy Broth without Dextrose

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

$200.18 - $200.18

Specifications

Certifications/Compliance BAM
Format Dehydrated
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Media Type Tryptic Soy Broth
Product Type Tryptic Soy broth without Dextrose
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DF0862-17-8

 
bd
B286220
Bottle 500g Each for $200.18
Description & Specifications

Specifications

Certifications/Compliance BAM
Format Dehydrated
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Media Type Tryptic Soy Broth
Product Type Tryptic Soy broth without Dextrose
Media Format Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L
Shelf Life 1825 Days
Storage Requirements 2-8°C

Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

1825 Days