Please login to your online account to display your discounted pricing

BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth


For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium

Manufacturer: bd  B249210

 View more versions of this product

Catalog No. DF0492-07-8

  • / Each
 

Description & Specifications

Specifications

Quantity 2kg
Packaging Pail
Format Dehydrated
Product Type Todd Hewitt broth
Media Type Todd Hewitt Broth
Storage Requirements Refrigerate
Shelf Life 1825 Days
For Use With (Application) Cultivation of beta-hemolytic streptococci, especially serological studies
Media Format Heart, infusion 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
Accessories

1825 Days