Please login to your online account to display your discounted pricing

BD Bacto™ Dehydrated Agar


Purified solidifying agent.  BD Difco&trade Bacto™ Dehydrated Agar is purified to remove extraneous matter. 

Manufacturer: bd  B214030

 View more versions of this product

Catalog No. DF0140-07-4

  • / Each
 

Description & Specifications

Specifications

Concentration 1.5%
Format Dehydrated
Packaging Container
Quantity 2000g
Certifications/Compliance ISO/DIS, 11014
For Use With (Application) Determination of motility and the growth of anaerobes and microaerophiles
Additive Agar
Shelf Life 1825 Days

  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling

Determines motility and growth of anaerobes and microaerophiles