Please login to your online account to display your discounted pricing

BD Bacto™ Casamino Acids


Suitable for use in diphtheria toxin production.   BD Difco™ Bacto™ Casamino Acids comprise acid hydrolyzed casein having low sodium chloride and iron concentrations.

Manufacturer: bd  B228830

 View more versions of this product

Catalog No. DF0288-17-4

  • / Each
 

Description & Specifications

Specifications

Format Dehydrated Powder
Additive Casamino Acid
Grade Vitamin Assay
Quantity 500g
Storage Requirements <30°C
Shelf Life 1825 Days
Packaging Bottle
Certifications/Compliance AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014
For Use With (Application) Vitamin assay

  • Dehydrated format
  • Peptones/ Hydrolysates
  • DNR (Latex) free

Accessories

1825 Days